siRNA Details
Gene Symbol: Itgb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177290.3 | NP_796264.2 | 2, 3 | 699 | ||
GenBank mRNA | AK029281.1 | - | 2, 3 | 1105 | ||
AK034644.1 | - | 2, 3 | 1194 | |||
AK053056.1 | - | 1224 | ||||
AK082444.1 | - | 1234 | ||||
BC120691.1 | - | 2, 3 | 699 | 40129614 | ||
BC125343.1 | - | 2, 3 | 699 | 40129858 | ||
BC144785.1 | - | 2, 3 | 699 |

More Information
Other Species Targeted

Gene Ontology Categories:
