siRNA Details
Gene Symbol: Dopey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177208.3 | NP_796182.2 | 5 | 730 | ||
GenBank mRNA | AK044820.1 | - | 4 | 620 | ||
AK045873.1 | - | 4 | 617 | |||
AK052544.1 | - | 4 | 619 | |||
AK054511.1 | - | 4 | 661 | |||
AK082462.1 | - | 4 | 653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
