siRNA Details
Gene Symbol: Pex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293806.1 | NP_001280735.1 | 8 | 1861 | ||
NM_027777.2 | NP_082053.1 | 7 | 1741 | |||
GenBank mRNA | AK082305.1 | - | 1 | 35 | ||
AK087687.1 | - | 3 | 299 | |||
BC090845.1 | - | 7 | 1741 | 6400629 | ||
DV646280.1 | - | 4 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
