siRNA Details
Gene Symbol: D5Ertd579e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081232.3 | NP_001074701.3 | 2, 3 | 670 | ||
GenBank mRNA | AK032804.1 | - | 2, 3 | 587 | ||
AK033480.1 | - | 2, 3 | 619 | |||
AK083736.1 | - | 2, 3 | 610 | |||
AK122224.1 | - | 1, 2 | 181 | |||
AK134363.1 | - | 2, 3 | 631 | |||
AK139370.1 | - | 2, 3 | 587 | |||
AK152987.1 | - | 2, 3 | 570 | |||
BC151085.1 | - | 1, 2 | 415 | 9087998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
