siRNA Details
Gene Symbol: Zscan5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133204.2 | NP_573467.2 | 2 | 172 | ||
GenBank mRNA | BC139442.1 | - | 1 | 62 | 8862464 | |
BC139456.1 | - | 1 | 62 | 8862478 | ||
CN717865.1 | - | 2 | 149 |
Gene Symbol: 6030443J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029857.1 | - | 3 | 674 | ||
AK031505.1 | - | 1 | 630 |
Gene Symbol: Armt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038060.1 | - | 3 | 1058 |
Gene Symbol: Fastkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083186.1 | - | 1651 |
Gene Symbol: Paqr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285845.1 | NP_001272774.1 | 3 | 2758 | ||
NM_001285846.1 | NP_001272775.1 | 4 | 2684 | |||
NM_001285847.1 | NP_001272776.1 | 3 | 2647 | |||
NM_001285849.1 | NP_001272778.1 | 3 | 2531 | |||
NM_027995.3 | NP_082271.1 | 2 | 2563 | |||
GenBank mRNA | AK161542.1 | - | 3 | 2405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
