siRNA Details
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133758.3 | NP_598519.2 | 3, 4 | 570 | ||
NM_177249.3 | NP_796223.2 | 4, 5 | 630 | |||
GenBank mRNA | AK041541.1 | - | 3, 4 | 550 | ||
AK082114.1 | - | 4, 5 | 594 | |||
AK083763.1 | - | 4, 5 | 847 | |||
AK151051.1 | - | 3, 4 | 574 | |||
BC108425.1 | - | 4, 5 | 571 | 30062566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
