siRNA Details
Gene Symbol: Dab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177259.4 | NP_796233.2 | 15 | 2482 | ||
GenBank mRNA | AK083206.1 | - | 2 | 546 | ||
BC138538.1 | - | 14 | 2203 | 8861558 | ||
BC138539.1 | - | 14 | 2204 | 8861559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
