siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 6 | 3362 | ||
GenBank mRNA | AK029850.1 | - | 1 | 436 | ||
AK030106.1 | - | 6 | 3364 | |||
AK042228.1 | - | 1 | 334 | |||
AK049629.1 | - | 1 | 572 | |||
AK050000.1 | - | 1 | 137 | |||
AK122469.1 | - | 1 | 1535 | |||
BC086807.1 | - | 1 | 1981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
