siRNA Details
Gene Symbol: Fsbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256142.1 | NP_001243071.1 | 1 | 341 | ||
GenBank mRNA | AK053419.1 | - | 1 | 340 | ||
AK147819.1 | - | 1 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
