siRNA Details
Gene Symbol: Itgb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177290.3 | NP_796264.2 | 3 | 712 | ||
GenBank mRNA | AK029281.1 | - | 3 | 1118 | ||
AK034644.1 | - | 3 | 1207 | |||
AK053056.1 | - | 1237 | ||||
AK082444.1 | - | 1247 | ||||
BC120691.1 | - | 3 | 712 | 40129614 | ||
BC125343.1 | - | 3 | 712 | 40129858 | ||
BC144785.1 | - | 3 | 712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
