siRNA Details
Gene Symbol: Cep250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129999.1 | NP_001123471.1 | 8 | 1131 | ||
NM_001130000.1 | NP_001123472.1 | 8 | 1126 | |||
NM_008383.3 | NP_032409.3 | 8 | 1131 | |||
NM_177217.3 | NP_796191.2 | 8 | 1126 | |||
GenBank mRNA | AK033281.1 | - | 8 | 1095 | ||
AK045557.1 | - | 8 | 1118 | |||
AK145562.1 | - | 8 | 1127 | |||
AK161443.1 | - | 8 | 1232 | |||
BC068153.1 | - | 8 | 1114 | |||
BC145268.1 | - | 7 | 953 | |||
BC150746.1 | - | 7 | 953 | 9087657 | ||
DQ148475.1 | - | 7 | 926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
