siRNA Details
Gene Symbol: D5Ertd579e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081232.3 | NP_001074701.3 | 3 | 698 | ||
GenBank mRNA | AK032804.1 | - | 3 | 615 | ||
AK033480.1 | - | 3 | 647 | |||
AK083736.1 | - | 3 | 638 | |||
AK122224.1 | - | 2 | 209 | |||
AK134363.1 | - | 3 | 659 | |||
AK139370.1 | - | 3 | 615 | |||
AK152987.1 | - | 3 | 598 | |||
BC151085.1 | - | 2 | 443 | 9087998 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109388, XM_006251140, XM_006251141.
Gene Ontology Categories:
