siRNA Details
Gene Symbol: Ncor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252313.1 | NP_001239242.1 | 3 | 335 | ||
NM_011308.3 | NP_035438.3 | 3 | 335 | |||
NM_177229.3 | NP_796203.1 | 4 | 520 | |||
GenBank mRNA | AK035813.1 | - | 4 | 512 | ||
AK077420.1 | - | 4 | 521 | |||
BC026623.1 | - | 4 | 706 | |||
BC029627.1 | - | 4 | 498 | |||
BC086657.1 | - | 3 | 301 | 30545994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
