siRNA Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271463.1 | NP_001258392.1 | 21 | 3113 | ||
NM_001271464.1 | NP_001258393.1 | 20 | 3032 | |||
NM_001271465.1 | NP_001258394.1 | 21 | 3140 | |||
NM_177266.5 | NP_796240.3 | 21 | 3146 | |||
GenBank mRNA | AK044371.1 | - | 20 | 3033 | ||
AK044750.1 | - | 1 | 162 | |||
AK052633.1 | - | 5 | 1457 | |||
AK083441.1 | - | 20 | 3152 | |||
BC057062.1 | - | 10 | 2924 |
Gene Symbol: Hexb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CD775639.1 | - | 1 | 86 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
