siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 9 | 4402 | ||
GenBank mRNA | AK029850.1 | - | 4 | 1476 | ||
AK049629.1 | - | 4 | 1621 | |||
AK050000.1 | - | 4 | 1186 | |||
AK122469.1 | - | 4 | 2584 | |||
AK164672.1 | - | 1 | 642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
