siRNA Details
Gene Symbol: Wscd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049685.1 | - | 2029 | |||
AK173026.1 | - | 1 | 2277 | |||
BC147280.1 | - | 1 | 1360 | 9056352 | ||
BC147281.2 | - | 1 | 1361 | 9056324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
