siRNA Details
Gene Symbol: Xpnpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177310.2 | NP_796284.1 | 9 | 2006 | ||
GenBank mRNA | AK088324.1 | - | 9 | 2006 | ||
BC137569.1 | - | 10 | 2168 | 8860588 | ||
BC144717.1 | - | 10 | 2169 | 9053265 |
