siRNA Details
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276292.1 | NP_001263221.1 | 6 | 601 | ||
NM_177327.5 | NP_796301.2 | 5 | 558 | |||
GenBank mRNA | AK032232.1 | - | 5 | 549 | ||
AK042432.1 | - | 5 | 549 | |||
AK137588.1 | - | 5 | 483 | |||
AK168567.1 | - | 5 | 509 | |||
BB662323.1 | - | 5 | 552 | |||
BC026829.1 | - | 6 | 539 | 4209397 | ||
BC150812.1 | - | 4 | 393 | 9087723 | ||
CJ137158.1 | - | 5 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
