siRNA Details
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035239.2 | NP_001030316.1 | 11, 12 | 1582 | ||
NM_001035240.2 | NP_001030317.1 | 11, 12 | 1582 | |||
NM_001035241.2 | NP_001030318.1 | 11, 12 | 1582 | |||
NM_001035242.1 | NP_001030319.1 | 11, 12 | 1582 | |||
NM_001035243.2 | NP_001030320.1 | 11, 12 | 1582 | |||
NM_177341.4 | NP_796315.2 | 11, 12 | 1649 | |||
GenBank mRNA | AJ544532.1 | - | 11, 12 | 1582 | ||
AJ544533.1 | - | 11, 12 | 1582 | |||
AJ544534.1 | - | 11, 12 | 1582 | |||
AJ544535.1 | - | 11, 12 | 1582 | |||
AJ544536.1 | - | 11, 12 | 1582 | |||
AK044454.1 | - | 11, 12 | 1651 | |||
AK051867.1 | - | 6, 7 | 715 | |||
AK142504.1 | - | 12, 13 | 1538 | |||
AK173218.1 | - | 11, 12 | 1653 | |||
BC141400.1 | - | 11, 12 | 1625 | 9056013 | ||
BC141412.1 | - | 11, 12 | 1625 | 9056025 | ||
HM064407.1 | - | 11, 12 | 1674 | |||
HM064408.1 | - | 11, 12 | 1674 | |||
HM064409.1 | - | 11, 12 | 1674 | |||
HM064410.1 | - | 12, 13 | 1749 | |||
JF706722.1 | - | 11, 12 | 1645 | |||
JX644970.1 | - | 11, 12 | 1674 | |||
JX644971.1 | - | 11, 12 | 1674 | |||
JX644972.1 | - | 11, 12 | 1674 | |||
JX644973.1 | - | 11, 12 | 1674 | |||
JX644974.1 | - | 11, 12 | 1674 | |||
JX644975.1 | - | 12, 13 | 1749 | |||
JX644979.1 | - | 12, 13 | 1749 | |||
JX644980.1 | - | 12, 13 | 1749 | |||
JX644981.1 | - | 12, 13 | 1749 | |||
JX644983.1 | - | 11, 12 | 1674 | |||
JX644984.1 | - | 11, 12 | 1674 | |||
JX644985.1 | - | 11, 12 | 1674 | |||
JX644986.1 | - | 11, 12 | 1674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
