siRNA Details
Gene Symbol: Ifna12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177361.2 | NP_796335.1 | 1 | 238 | ||
GenBank mRNA | AY190046.1 | - | 1 | 238 | ||
BC104352.1 | - | 1 | 217 | 40049569 | ||
BC104353.1 | - | 1 | 217 | 40049571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
