siRNA Details
Gene Symbol: Sfmbt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198808.1 | NP_001185737.1 | 3, 4 | 305 | ||
NM_001198809.1 | NP_001185738.1 | 3, 4 | 384 | |||
NM_177386.5 | NP_796360.2 | 4, 5 | 428 | |||
GenBank mRNA | AK040600.1 | - | 3, 4 | 385 | ||
AK049505.1 | - | 3, 4 | 247 | |||
AK052339.1 | - | 3, 4 | 371 | |||
AK147557.1 | - | 3, 4 | 310 | |||
AK147651.1 | - | 4, 5 | 283 | |||
AK168444.1 | - | 5, 6 | 333 | |||
AK220408.1 | - | 1, 2 | 170 | |||
BB636293.1 | - | 3, 4 | 385 | |||
BC115510.1 | - | 3, 4 | 239 | |||
BC115511.1 | - | 3, 4 | 239 | |||
BC139354.1 | - | 3, 4 | 216 | 8862376 | ||
BC145610.1 | - | 2, 3 | 172 | 9054176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
