siRNA Details
Gene Symbol: Tmem191c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177473.3 | NP_803424.2 | 2, 3 | 295 | ||
GenBank mRNA | AK016675.1 | - | 2, 3 | 282 | ||
BU936413.1 | - | 2, 3 | 278 | |||
BY296351.1 | - | 2, 3 | 150 |

More Information
Other Species Targeted

Gene Ontology Categories:
