siRNA Details
Gene Symbol: Gm13088 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126325.1 | NP_001119797.1 | 2 | 156 |
Gene Symbol: Gm13103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177571.3 | NP_808239.2 | 2 | 164 | ||
GenBank mRNA | AK054347.1 | - | 2 | 165 | ||
BC096438.1 | - | 2 | 161 | 30481183 | ||
BY029530.1 | - | 2 | 165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
