siRNA Details
Gene Symbol: Dnah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033668.1 | NP_001028840.1 | 71, 72 | 11461 | ||
GenBank mRNA | AK046346.1 | - | 13, 14 | 2086 | ||
AK052631.1 | - | 13, 14 | 2088 | |||
AK163552.1 | - | 13, 14 | 2086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
