siRNA Details
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035240.2 | NP_001030317.1 | 12, 13 | 1635 | ||
NM_001035241.2 | NP_001030318.1 | 12, 13 | 1635 | |||
NM_177341.4 | NP_796315.2 | 12, 13 | 1702 | |||
GenBank mRNA | AJ544533.1 | - | 12, 13 | 1635 | ||
AJ544534.1 | - | 12, 13 | 1635 | |||
AK044454.1 | - | 12, 13 | 1704 | |||
AK051867.1 | - | 7, 8 | 768 | |||
BC141400.1 | - | 12, 13 | 1678 | 9056013 | ||
BC141412.1 | - | 12, 13 | 1678 | 9056025 | ||
HM064409.1 | - | 12, 13 | 1727 | |||
JF706722.1 | - | 12, 13 | 1698 | |||
JX644972.1 | - | 12, 13 | 1727 | |||
JX644973.1 | - | 12, 13 | 1727 | |||
JX644977.1 | - | 12, 13 | 1673 | |||
JX644978.1 | - | 12, 13 | 1673 | |||
JX644979.1 | - | 13, 14 | 1802 | |||
JX644980.1 | - | 13, 14 | 1802 | |||
JX644982.1 | - | 13, 14 | 1748 | |||
JX644983.1 | - | 12, 13 | 1727 | |||
JX644984.1 | - | 12, 13 | 1727 | |||
JX644985.1 | - | 12, 13 | 1727 | |||
JX644986.1 | - | 12, 13 | 1727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
