siRNA Details
Gene Symbol: Zfp799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293229.1 | NP_001280158.1 | 2 | 291 | ||
NM_177359.5 | NP_796333.3 | 2 | 291 | |||
GenBank mRNA | AK031686.1 | - | 2 | 279 | ||
AK083640.1 | - | 2 | 268 | |||
AK137037.1 | - | 2 | 268 | |||
BC062813.1 | - | 2 | 245 | |||
BC096552.1 | - | 2 | 241 | 30618827 |
