siRNA Details
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175275.4 | NP_780484.2 | 2 | 533 | ||
NM_177385.4 | NP_796359.1 | 2 | 533 | |||
GenBank mRNA | AK031604.1 | - | 2 | 439 | ||
AK052558.1 | - | 2 | 453 | |||
BC125622.1 | - | 2 | 408 | |||
BC125624.1 | - | 2 | 408 | |||
BM942195.1 | - | 2 | 533 |
