siRNA Details
Gene Symbol: Sfmbt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198808.1 | NP_001185737.1 | 4 | 323 | ||
NM_001198809.1 | NP_001185738.1 | 4 | 402 | |||
NM_177386.5 | NP_796360.2 | 5 | 446 | |||
GenBank mRNA | AK040600.1 | - | 4 | 403 | ||
AK049505.1 | - | 4 | 265 | |||
AK052339.1 | - | 4 | 389 | |||
AK147557.1 | - | 4 | 328 | |||
AK147651.1 | - | 5 | 301 | |||
AK168444.1 | - | 6 | 351 | |||
AK220408.1 | - | 2 | 188 | |||
BB636293.1 | - | 4 | 403 | |||
BC115510.1 | - | 4 | 257 | |||
BC115511.1 | - | 4 | 257 | |||
BC139354.1 | - | 4 | 234 | 8862376 | ||
BC145610.1 | - | 3 | 190 | 9054176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
