siRNA Details
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177465.4 | NP_803416.2 | 18 | 3603 | ||
GenBank mRNA | AK040446.1 | - | 10 | 2104 | ||
AY771616.1 | - | 19 | 3520 | |||
AY771617.1 | - | 18 | 3607 | |||
AY771618.1 | - | 18 | 3262 | |||
BC051427.1 | - | 18 | 3258 | |||
BC132381.1 | - | 5 | 792 |
