siRNA Details
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035239.2 | NP_001030316.1 | 25 | 5852 | ||
NM_001035240.2 | NP_001030317.1 | 26 | 5852 | |||
NM_001035241.2 | NP_001030318.1 | 25 | 5822 | |||
NM_001035242.1 | NP_001030319.1 | 25 | 5816 | |||
NM_001035243.2 | NP_001030320.1 | 24 | 5786 | |||
NM_177341.4 | NP_796315.2 | 27 | 4265 | |||
GenBank mRNA | AK051867.1 | - | 22 | 3331 | ||
AK142504.1 | - | 26 | 5772 | |||
AK173218.1 | - | 25 | 5887 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
