siRNA Details
Gene Symbol: Setx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198033.2 | NP_932150.2 | 10 | 2953 | ||
GenBank mRNA | AK044730.1 | - | 10 | 2955 | ||
AK048354.1 | - | 1 | 2108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
