siRNA Details
Gene Symbol: Sfmbt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198809.1 | NP_001185738.1 | 6 | 1418 | ||
GenBank mRNA | AK040600.1 | - | 6 | 1419 | ||
AK049505.1 | - | 6 | 1281 | |||
AK168444.1 | - | 8 | 1368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
