siRNA Details
Gene Symbol: Igsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177591.4 | NP_808259.2 | 12 | 1993 | ||
NM_183335.2 | NP_899178.2 | 11 | 1857 | |||
NM_183336.2 | NP_899179.1 | 3 | 247 | |||
GenBank mRNA | AK030701.1 | - | 3 | 249 | ||
AK076427.1 | - | 3 | 240 | |||
AK129126.1 | - | 12 | 1975 | |||
AY227771.1 | - | 12 | 1987 | |||
AY227773.1 | - | 11 | 1857 | |||
AY227774.1 | - | 3 | 242 | |||
BC119264.1 | - | 3 | 210 | 8734013 | ||
BC119266.1 | - | 3 | 210 | 8734015 | ||
BC144762.1 | - | 3 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
