siRNA Details
Gene Symbol: Unc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081017.2 | NP_001074486.2 | 39, 40 | 7227 | ||
GenBank mRNA | AB093296.1 | - | 22, 23 | 4315 | ||
AB257853.1 | - | 39, 40 | 7227 | |||
AK033439.1 | - | 2, 3 | 198 | |||
GQ334471.1 | - | 42, 43 | 6706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
