siRNA Details
Gene Symbol: Zmym2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029498.3 | NP_083774.2 | 2 | 361 | ||
GenBank mRNA | AK135499.1 | - | 3 | 474 | ||
BC046507.1 | - | 3 | 407 | |||
BC141403.1 | - | 2 | 337 | 9056016 | ||
BU055093.1 | - | 3 | 475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
