siRNA Details
Gene Symbol: Dgkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177646.3 | NP_808314.2 | 23, 24 | 2856 | ||
GenBank mRNA | AK172901.1 | - | 19, 20 | 2301 | ||
BC048957.1 | - | 2, 3 | 312 | |||
BC057061.1 | - | 18 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
