siRNA Details
Gene Symbol: Cstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177655.3 | NP_808323.1 | 2 | 236 | ||
GenBank mRNA | AK131797.1 | - | 2 | 237 | ||
AK132807.1 | - | 2 | 239 | |||
AY707915.1 | - | 2 | 236 | |||
BC048646.1 | - | 2 | 236 | 6772392 |

More Information
Other Species Targeted

Gene Ontology Categories:
