siRNA Details
Gene Symbol: 6820408C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289738.1 | NP_001276667.1 | 5 | 610 | ||
NM_177656.4 | NP_808324.1 | 5 | 607 | |||
GenBank mRNA | AK078482.1 | - | 5 | 607 | ||
BC137826.1 | - | 4 | 487 | 8860845 | ||
BC144943.1 | - | 4 | 484 | |||
BC144944.1 | - | 4 | 487 | 9053492 | ||
BC144945.1 | - | 4 | 480 |
