siRNA Details
Gene Symbol: Ralgapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291137.1 | NP_001278066.1 | 30 | 4550 | ||
NM_001291138.1 | NP_001278067.1 | 29 | 4514 | |||
GenBank mRNA | AK047718.1 | - | 5 | 530 | ||
AK122470.1 | - | 25 | 3264 | |||
BC138377.1 | - | 30 | 4452 | 8861397 | ||
BC145260.1 | - | 29 | 4417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
