siRNA Details
Gene Symbol: Zbtb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177660.3 | NP_808328.2 | 5 | 2748 | ||
GenBank mRNA | AK048280.1 | - | 4 | 1210 | ||
AK083443.1 | - | 3 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
