siRNA Details
Gene Symbol: Igsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177591.4 | NP_808259.2 | 13 | 2246 | ||
NM_183335.2 | NP_899178.2 | 12 | 2110 | |||
NM_183336.2 | NP_899179.1 | 4 | 500 | |||
GenBank mRNA | AK030701.1 | - | 4 | 502 | ||
AK076427.1 | - | 4 | 493 | |||
AK129126.1 | - | 13 | 2228 | |||
AY227771.1 | - | 13 | 2240 | |||
AY227773.1 | - | 12 | 2110 | |||
AY227774.1 | - | 4 | 495 | |||
BC119264.1 | - | 4 | 463 | 8734013 | ||
BC119266.1 | - | 4 | 463 | 8734015 | ||
BC144762.1 | - | 4 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
