siRNA Details
Gene Symbol: Fbxw13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177598.3 | NP_808266.1 | 4 | 362 | ||
GenBank mRNA | AK054339.1 | - | 4 | 363 | ||
BC119542.1 | - | 4 | 349 | 40056515 | ||
BC127237.1 | - | 3 | 185 | 40056507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
