siRNA Details
Gene Symbol: 4932415M13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030017.1 | - | 2 | 781 | ||
BC109354.1 | - | 1 | 598 | 40057021 | ||
BC116728.1 | - | 1 | 598 | 40106470 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4932415M13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030017.1 | - | 2 | 781 | ||
BC109354.1 | - | 1 | 598 | 40057021 | ||
BC116728.1 | - | 1 | 598 | 40106470 |