siRNA Details
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177608.3 | NP_808276.2 | 4 | 826 | ||
GenBank mRNA | AK049924.1 | - | 4 | 827 | ||
AK145135.1 | - | 4 | 712 | |||
AK169713.1 | - | 4 | 713 | |||
AK172920.1 | - | 4 | 695 | |||
BC095939.1 | - | 4 | 693 | |||
BC141056.1 | - | 4 | 786 | 9055668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
