siRNA Details
Gene Symbol: Unc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081017.2 | NP_001074486.2 | 40, 41 | 7339 | ||
GenBank mRNA | AB093296.1 | - | 23, 24 | 4427 | ||
AB257853.1 | - | 40, 41 | 7339 | |||
AK033439.1 | - | 3, 4 | 310 | |||
GQ334471.1 | - | 43, 44 | 6818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
