siRNA Details
Gene Symbol: Ccdc148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001178.1 | NP_001001178.1 | 1849 | |||
GenBank mRNA | AK030792.1 | - | 4 | 357 | ||
BC062650.1 | - | 1849 | 6417188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
