siRNA Details
Gene Symbol: Ralgapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291137.1 | NP_001278066.1 | 31 | 4632 | ||
NM_001291138.1 | NP_001278067.1 | 30 | 4596 | |||
GenBank mRNA | AK047718.1 | - | 6 | 612 | ||
AK122470.1 | - | 26 | 3346 | |||
BC138377.1 | - | 31 | 4534 | 8861397 | ||
BC145260.1 | - | 30 | 4499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
