siRNA Details
Gene Symbol: Tmem201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284273.1 | NP_001271202.1 | 6 | 1891 | ||
NM_177672.4 | NP_808340.2 | 6 | 1904 | |||
GenBank mRNA | AK040555.1 | - | 3 | 2329 | ||
AK079157.1 | - | 6 | 1811 | |||
AK086543.1 | - | 6 | 1886 | |||
AK153631.1 | - | 6 | 1881 | |||
AK159041.1 | - | 6 | 1892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
