siRNA Details
Gene Symbol: Cfap74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177674.5 | NP_808342.3 | 8, 9 | 1170 | ||
GenBank mRNA | AK040922.1 | - | 3, 4 | 679 | ||
AK134858.1 | - | 8, 9 | 1102 |

More Information
Other Species Targeted

Gene Ontology Categories:
