siRNA Details
Gene Symbol: Thnsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177588.2 | NP_808256.1 | 3 | 1038 | ||
GenBank mRNA | AK029518.1 | - | 3 | 1039 | ||
AK083895.1 | - | 3 | 1038 | |||
BC090667.1 | - | 1 | 747 | 30686917 | ||
BC117854.2 | - | 2 | 822 | 40094242 |

More Information
Other Species Targeted

Gene Ontology Categories:
